Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL-1 beta Monoclonal Antibody (ILB1-H6), Biotin
GREENER_CHOICE

Catalog No. ENM420BB Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ENM420BB 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ENM420BB Supplier Invitrogen™ Supplier No. M420BB
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

M420BB targets IL-1 beta in ELISA applications and shows reactivity with Human samples. The M420BB immunogen is recombinant human IL-1 beta. M420BB detects IL-1 beta which has a predicted molecular weight of approximately 31 kDa. The M420BB IL1b antibody (biotinylated conjugate of clone ILB1-H6) has successfully been paired as the detection antibody in a sandwich ELISA with capture antibody M421B (clone ILB1-H67). Typical dilutions for sandwich ELISA include 1 μg/mL for coating and 0.125 - 0.25 μg/mL for detection. Biotinylated antibody M420BB (clone ILB1-H6) and antibody M421B (clone ILB1-H67) have successfully been used in combination with recombinant IL1 beta protein SIL1B in ELISA applications.

Interleukin-1 beta (IL-1 beta) is a proinflammatory cytokine expressed by monocytes, macrophages, and dendritic cells. IL-1 beta is synthesized in response to inflammatory stimuli as a 31 kDa inactive pro-form that accumulates in the cytosol. Cleavage of pro-IL-1 beta into the active 17 kDa protein requires the activation of inflammasomes, which are multi-protein complexes that respond to pathogens, stress conditions, and other danger signals. Inflammasome activation triggers the processing of the caspase-1 precursor into its active form, which in turn cleaves pro-IL-1 beta. IL-1 beta lacks a signal sequence peptide for classical ER/Golgi pathway and is secreted alongside caspase-1 via an alternate and incompletely understood mechanism. Although IL-1 beta is most often secreted in its active form, secretion of the uncleaved protein may be detectable under some biological conditions. IL-1 beta signals through two receptors, IL-1RI and IL-1RII, both of which are shared with IL-1 alpha. IL-1 beta activity can be moderated by IL-1 Receptor Antagonist (IL-1RA), a protein produced by many cell types that blocks receptor binding through competitive inhibition. IL-1 beta play an important role in innate host defense by triggering the production of other proinflammatory cytokines in target cells and initiating acute-phase responses to infection and injury. Elevated levels of IL-1 beta have been associated with many chronic inflammatory conditions IL-1 beta neutralizing antibodies potential therapeutic value.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-1 beta
Applications ELISA
Classification Monoclonal
Clone ILB1-H6
Concentration 0.5 mg/mL
Conjugate Biotin
Formulation PBS with 4% BSA and proprietary Preservative
Gene IL1B
Gene Accession No. P01584
Gene Alias catabolin; cytokine; Hematopoietin 1 (H1); IFN beta inducing factor; il 1b; IL 1β; IL-1; IL1 B; IL-1 beta; IL-1 precursor; IL1B; IL-1B; IL1B1; IL1beta; IL-1beta; IL1-BETA; IL1F2; IL1β; ILN; Interleukin; interleukin 1 beta; Interleukin 1 beta precursor; interleukin 1, beta; interleukin 1, beta 1; interleukin 1-beta; interleukin*1*beta; Interleukin1 beta; interleukin-1 beta; interleukin-1 beta precursor; interleukin-1 beta precursor (AA -113 to 153); interleukin-1 beta proprotein; interleukin-1b; interleukin-1beta; interleukin-beta; LAF; lymphocyte proliferation-potentiating factor; Osteoclast activating factor (OAF); preinterleukin 1 beta; Pro interleukin 1 beta; prointerleukin-1 beta; pro-interleukin-1-beta
Gene Symbols IL1B
Host Species Mouse
Immunogen Recombinant human IL-1 beta.
Purification Method Protein G
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3553
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.