Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ IL-1 RI Antibody Pair [HRP]
SDP

Catalog No. NB010289 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
15 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB010289 15 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB010289 Supplier Novus Biologicals™ Supplier No. NBP27962515PLATES

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody Pair

specifically detects in Rat samples. It is validated for ELISA, Sandwich ELISA.

Specifications

Assay Range 62.5 pg/mL
Assay Sensitivity 62.5-4000 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Rat IL-1 RI.
Target Species Rat
For Use With (Application) Sandwich ELISA
Gene Alias CD121 antigen-like family member A, CD121a antigen, IL1R, IL1R1, IL1RT1, IL-1RT1, IL-1RT-1, Interleukin 1 Receptor 1, interleukin 1 receptor, type I, interleukin receptor 1, interleukin-1 receptor type 1, p80
Gene ID (Entrez) 3554
Gene Symbol IL1R1
Host Species Mouse
Kit Contents
  • Detection Antibody: 0.25 mg/mL mouse anti-IL-1 RI monoclonal antibody conjugated to horseradish-peroxidase (HRP).
  • Capture Antibody: 1.0 mg/mL of mouse anti-rat IL-1 RI monoclonal antibody.
  • Standard: Each vial contains 148 ng of recombinant rat IL-1 RI.
  • Quantity 15 Plates
    Regulatory Status RUO
    Research Discipline Cancer, Cytokine Research, Immunology, Innate Immunity, Phospho Specific
    Storage Requirements Storage is content dependent.
    Target IL-1 RI
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 4 ng/mL is recommended.
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.