Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL-10 Monoclonal Antibody (JES5-16E3), FITC, eBioscience™
GREENER_CHOICE

Catalog No. 11710141
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-710-141 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-710-141 Supplier Invitrogen™ Supplier No. 11710141
  • $113.00
    $84.65 / Each of 1

    Save $28.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rat Monoclonal Antibody

Description: The JES5-16E3 antibody reacts with mouse interleukin-10 (IL-10). Mouse IL-10 is an ~18 kDa factor also known as Cytokine Synthesis Inhibitory Factor (CSIF). In the mouse, Th2 cells, B1 cells, macrophages, and keratinocytes are the major cell subsets that produce IL-10. IL-10 inhibits synthesis of Th1 cytokines and proliferation of T cells, and acts as a costimulatory signal for mast cells, developing thymocytes and the Th2 response. Applications Reported: The JES5-16E3 antibody has been reported for use in capture of mouse IL-10 by ELISA and ELISPOT, intracellular staining for flow cytometric analysis, IHC, and neutralization of IL-10 bioactivity. Applications Tested: This JES5-16E3 antibody has been pre-titrated and tested by intracellular staining for flow cytometric analysis. test size: has been pre-titrated and tested by intracellular staining and flow cytometric analysis of LPS-stimulated normal human peripheral blood cells. This can be used at test size: 5 μL (0.25 μg)/test per test. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.

IL-10 encodes a protein that acts as a cytokine and is primarily produced by monocytes, with some production by lymphocytes. This cytokine has various effects on immunoregulation and inflammation, including down-regulating the expression of Th1 cytokines, MHC class II Ags, and costimulatory molecules on macrophages. It also enhances B cell survival, proliferation, and antibody production. Additionally, IL-10 can block NF-kappa B activity and is involved in the regulation of the JAK-STAT signaling pathway. Knockout studies in mice have suggested that this cytokine is an essential immunoregulator in the intestinal tract. Mutations in this gene have been associated with an increased susceptibility to HIV-1 infection and rheumatoid arthritis. Diseases associated with IL10 include Graft-Versus-Host Disease and Human Immunodeficiency Virus Type 1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-10
Applications Flow Cytometry
Classification Monoclonal
Clone JES5-16E3
Concentration 5 μL/Test
Conjugate FITC
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene IL10
Gene Accession No. P18893
Gene Alias CSIF; cytokine; Cytokine synthesis inhibitory factor; GVHDS; H-IL-10; il 10; IL10; IL-10; IL-10 precursor; IL10A; IL10X; ILN; Interleukin; interleukin 10; Interleukin10; interleukin-10; MGC126450; MGC126451; RP11-262N9.1; T-cell growth inhibitory factor; TGIF
Gene Symbols IL10
Host Species Rat
Purification Method Affinity chromatography
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16153
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.