Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL-11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA536544 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA536544 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA536544 Supplier Invitrogen™ Supplier No. PA536544
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

Purity is >95% by SDS-PAGE.

Interleukins (ILs) are a large group of cytokines that are produced mainly by leukocytes, although some are produced by certain phagocytes and auxiliary cells. ILs have a variety of functions, but most function to direct other immune cells to divide and differentiate. Each IL acts on a specific, limited group of cells through a receptor specific for that IL.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-11
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene IL11
Gene Accession No. P20809, P47873, Q99MF5
Gene Alias adipogenesis inhibitory factor; AGIF; il 11; IL10RA; Il11; IL-11; IL11 inhibitor; IL-11ra; ILN; Interleukin; interleukin 11; Interleukin11; interleukin-11; MGC2146; Oprelvekin
Gene Symbols IL11
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the N-terminus of human IL-11.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16156, 171040, 3589
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.