Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL-12/IL-23 p40 Monoclonal Antibody (C8.6), Brilliant Ultra Violet™ 395, eBioscience™

Catalog No. 363712941
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-371-2941 25 Tests
36-371-2942 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 36-371-2941 Supplier Invitrogen™ Supplier No. 363712941
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The C8.6 antibody reacts with the p40 subunit of human IL-12 p70 and human IL-23. IL-12 is a 70 kDa heterodimeric cytokine composed of two covalently linked chains, p40 and p35, and secreted primarily by activated monocytes, macrophages, and dendritic cells. IL-23 is a biologically-related, heterodimeric cytokine composed of disulfide-linked p40 and p19 subunits. Applications Tested This C8.6 antibody has been pre-diluted and tested by intracellular staining followed by flow cytometric analysis of normal human peripheral blood cells using the Intracellular Fixation & Permeabilization Buffer Set (Product # 88-8824-00) and protocol. Please refer to 'Staining Intracellular Antigens for Flow Cytometry, Protocol A: Two step protocol for intracellular (cytoplasmic) proteins' located at Flow Protocols. This may be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Blocking Buffers When using two or more Super Bright, Brilliant Violet™, Brilliant Ultra Violet™, or other polymer dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Complete Staining Buffer (Product # SB-4401) or Brilliant Stain Buffer (Product # 00-4409-75) to minimize any non-specific polymer interactions. Please refer to the datashee...

IL-12 is an interleukin produced by dendritic cells, neutrophils, macrophages and human lymphoblastoid cells as a result of antigenic stimulation. IL-12 is a heterodimeric cytokine encoded by IL-12A (p35) and IL-12B (p40). IL-23 is a heterodimeric cytokine that shares a subunit with IL-12 (IL-12B) along with an IL-23A (IL-23p19) subunit. Functionally, IL-12 increases the expression of vascular endothelial adhesion molecule-1 on the endothelium, directs leukocytes towards tumors and facilitates the ischemic-hemorrhagic necrosis of the tumor tissue. In comparison, IL-23 plays an important role in stimulating memory T-cells, and is required for the induction, expansion, maintenance and downstream effector functions of Th17 cells, which play a vital role in upregulating neutrophil chemokines and various pro-inflammatory cytokines.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-12/IL-23 p40
Applications Flow Cytometry
Classification Monoclonal
Clone C8.6
Concentration 5 μL/Test
Conjugate Brilliant Ultraviolet 395
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene IL12A
Gene Accession No. P29459, P29460, Q9NPF7
Gene Alias CLMF; CLMF p35; CLMF p40; CLMF2; cytotoxic lymphocyte maturation factor 1, p35; cytotoxic lymphocyte maturation factor 35 kDa subunit; Cytotoxic lymphocyte maturation factor 40 kDa subunit; il 12; IL 12/IL 23p40; il 23; IL 23 p40; Il12; Il-12 p35; IL12 p40; IL23; IL-12 subunit p35; IL-12 subunit p40; IL-12, subunit p35; IL12, subunit p40; Il12a; IL-12A; Il12b; IL-12B; IL12p35; IL-12p35; Il12p40; Il-12p40; il23; IL-23; IL23 p41; IL-23 subunit alpha; Il23a; IL-23A; IL-23-A; IL23P19; IL-23p19; IL35 subunit; ILN; IMD28; IMD29; Interleukin; Interleukin 12; interleukin 12 35 kDa subunit; interleukin 12 35kDa subunit; interleukin 12 40 kDa subunit; interleukin 12 40kDa subunit; interleukin 12 p35 subunit; interleukin 12 polypeptide 35; interleukin 12 subunit p40; interleukin 12, p35; interleukin 12, p40; interleukin 12A; interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocyte maturation factor 1, p35); interleukin 12b; interleukin 12B (natural killer cell stimulatory factor 2, cytotoxic lymphocyte maturation factor 2, p40); interleukin 23 p19 subunit; interleukin 23 subunit alpha; interleukin 23, alpha subunit p19; Interleukin12; interleukin-12 alpha chain; interleukin-12 beta chain; interleukin-12 p40; interleukin-12 p40 subunit; interleukin-12 subunit alpha; Interleukin-12 subunit beta; interleukin-12 subunit p40; interleukin-23 p19 subunit; interleukin-23 p40 subunit; interleukin-23 subunit alpha; Interleukin-23 subunit p19; interleukin-six, G-CSF related factor; JKA3 induced upon T-cell activation; Ll12a; natural killer cell stimulatory factor 1, cytotoxic lymphocyte maturation factor 1, p35; natural killer cell stimulatory factor, 40 kD subunit; NF cell stimulatory factor chain 1; NFSK; NK cell stimulatory factor chain 1; NK cell stimulatory factor chain 2; NKSF; NKSF1; NKSF2; p19; P35; p40; RP23-388G23.1; SGRF; UNQ2498/PRO5798
Gene Symbols IL12A, IL12B, Il23a
Host Species Mouse
Purification Method Affinity chromatography
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3592, 3593, 51561
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.