Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL-12 p35 Monoclonal Antibody (SNKY35), eFluor™ 660, eBioscience™
GREENER_CHOICE

Catalog No. 50735942
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-735-942 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-735-942 Supplier Invitrogen™ Supplier No. 50735942
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Description: This SNKY35 monoclonal antibody reacts with human p35, a component of two of the four members of the interleukin (IL)-12 family of cytokines. The IL-12 family members are heterodimeric cytokines composed of an alpha-chain, such as p35, and a beta-chain, such as p40. The alpha-chain proteins are thought to be the limiting factors for secretion of IL-12 family member cytokines, as expression of the alpha-chain is more restricted than that of the beta-chain proteins, and the two subunits may need to be co-expressed in order for cytokine secretion to occur. IL-12 family members are secreted primarily by antigen-presenting cells in response to various TLR agonists and other inflammatory stimuli. The exact types of stimuli present and kinetics of stimulation determine the balance of IL-12 family member production. The p35 protein was first identified as part of the IL-12 cytokine, which is also known as IL-12p70. Secretion is induced by IFN gamma and agonists of TLR3, 4, or 7 and is inhibited by Th2 cytokines. IL-12 is essential for the differentiation and maintenance of Th1 cells. The other subunit, p40, is expressed in much higher quantities than p35, and may be secreted independently as a monomer or homodimer. Free p40 may act as an antagonist of IL-12 function. More recently, p35 was also found to dimerize with EBI3 (Epstein-Barr Virus-Induced Gene 3) to form IL-35. The biology of IL-35 is incompletely understood, and it is believed to be unique among the IL-12...

Interleukin 12A is a novel signaling cytokine belonging to the Interleukin-6 protein superfamily. Interleukin-12 alpha chain precursor (IL-12A) is a 35-kD subunit of disulfide-linked heterodimer of cytokine IL-12 and has a broad array of biological activities. Reports suggest that IL-12A can act as a growth factor for activated T and NK cells and enhance the lytic activity of NK/lymphokine-activated Killer cells. IL-12A stimulates the production of IFN-gamma by resting PBMC as well as T-cell-independent induction of interferon (IFN)-gamma, and helps in the differentiation of both Th1 and Th2 cells. It is also involved in positive regulation of tyrosine phosphorylation of Stat4 protein. The signaling process of IL-12A in innate immunity requires nitric oxide synthase 2A. Along with EBI3, IL-12A forms a novel heterodimeric cytokine, designated as interleukin-35 (IL-35), that may be specifically produced by T reg cells and is required for maximal suppressive activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-12 p35
Applications Flow Cytometry
Classification Monoclonal
Clone SNKY35
Concentration 5 μL/Test
Conjugate eFluor 660
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene IL12A
Gene Accession No. P29459
Gene Alias CLMF; CLMF p35; cytotoxic lymphocyte maturation factor 1, p35; cytotoxic lymphocyte maturation factor 35 kDa subunit; il 12; IL-12 subunit p35; IL-12, subunit p35; Il12a; IL-12A; IL12p35; IL-12p35; IL35 subunit; ILN; Interleukin; Interleukin 12; interleukin 12 p35 subunit; interleukin 12, p35; interleukin 12a; interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocyte maturation factor 1, p35); Interleukin12; interleukin-12 alpha chain; interleukin-12 beta p35 subunit; interleukin-12 p35 subunit; interleukin-12 subunit alpha; Ll12a; NF cell stimulatory factor chain 1; NFSK; NK cell stimulatory factor chain 1; NKSF1; P35
Gene Symbols IL12A
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3592
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.