Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL-12 p70 Monoclonal Antibody (20C2)

Catalog No. ENM122
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ENM122 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ENM122 Supplier Invitrogen™ Supplier No. M122
Only null left
Add to Cart
Edge
Add to Cart

Rat Monoclonal Antibody

M122 targets IL-12 in ELISA, and WB applications and shows reactivity with Human samples. The M122 immunogen is recombinant human IL-12 (p70). M122 detects IL-12 which has a predicted molecular weight of approximately 23 kDa. The M122 IL12 antibody (clone 20C2) has successfully been paired as the coating antibody in a sandwich ELISA with detection antibody M121B (biotinylated conjugate of clone C8.6) Typical dilutions for sandwich ELISA include 1 μg/mL for coating and 0.125 - 0.25 μg/mL for detection. Antibody M122 (clone 20C2) and biotinylated antibody M121B (clone C8.6) have successfully been used in combination with recombinant IL12 p70 protein SIL12 in ELISA applications. This product has been tested for endotoxins by limulus amoebocyte lysate (LAL) assay and contains an endotoxin concentration of less than or equal to 10 endotoxin units per milligram (EU/mg).

IL-12 p70, also known as interleukin-12, is a cytokine that plays a critical role in the immune system's response to infections and cancer. It is produced by immune cells, including dendritic cells, macrophages, and B cells, in response to certain pathogens and tumor cells. IL-12 p70 stimulates the production of interferon-gamma and other cytokines, which promote the activation and proliferation of T cells and natural killer cells. IL-12 p70 has been studied as a potential therapeutic target for various diseases and conditions, including infectious diseases, autoimmune disorders, and cancer. It has been shown to enhance immune responses against certain infections and tumors, and to modulate inflammatory responses in autoimmune diseases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-12 p70
Applications ELISA, Western Blot
Classification Monoclonal
Clone 20C2
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene IL12A
Gene Accession No. P29459, P29460
Gene Alias 20C2; CLMF; CLMF p35; CLMF p40; CLMF2; cytokine; cytotoxic lymphocyte maturation factor 1, p35; cytotoxic lymphocyte maturation factor 35 kDa subunit; Cytotoxic lymphocyte maturation factor 40 kDa subunit; il 12; IL 12p70; Il12; IL-12; Il-12 p35; IL-12 p75; IL-12 subunit p35; IL-12 subunit p40; IL-12, subunit p35; IL12, subunit p40; Il-12; cytokine; IL12A; IL-12A; IL12A/B; IL12AB; IL12B; Il-12b; IL12P35; IL-12p35; Il12p40; Il-12p40; IL35 subunit; ILN; IMD28; IMD29; Interleukin; interleukin 12 35 kDa subunit; interleukin 12 35kDa subunit; interleukin 12 40 kDa subunit; interleukin 12 40kDa subunit; interleukin 12 p35 subunit; interleukin 12 p40 subunit precursor; interleukin 12 polypeptide 35; interleukin 12, p35; interleukin 12, p40; interleukin 12A; interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocyte maturation factor 1, p35); interleukin 12B; interleukin 12B (natural killer cell stimulatory factor 2, cytotoxic lymphocyte maturation factor 2, p40); interleukin 12p35 subunit precursor; interleukin-12 alpha chain; interleukin-12 alpha p40 subunit; interleukin-12 beta chain; interleukin-12 beta p35 subunit; interleukin-12 p35 subunit; interleukin-12 p35 subunit precursor; interleukin-12 p40; interleukin-12 p40 subunit; interleukin-12 p40 subunit precursor; Interleukin-12 p70; interleukin-12 subunit alpha; Interleukin-12 subunit beta; interleukin-23 p40 subunit; Ll12a; MGC151228; MGC151232; natural killer cell stimulatory factor 1, cytotoxic lymphocyte maturation factor 1, p35; natural killer cell stimulatory factor, 40 kD subunit; NF cell stimulatory factor chain 1; NFSK; NK cell stimulatory factor chain 1; NK cell stimulatory factor chain 2; NKSF; NKSF1; NKSF2; P35; p40
Gene Symbols IL12A, Il12b
Host Species Rat
Immunogen Recombinant human IL-12 (p70).
Purification Method Protein G
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3592, 3593
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.