Learn More
ACROBiosystems ACROBiosystems Biotinylated Human IL-12 R beta 1 / CD212 Protein, Fc,Avitag™
Supplier:  ACROBiosystems ILBH82F7200UG
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human IL-12 R beta 1, Fc,Avitag (ILB-H82F7) is expressed from human 293 cells (HEK293). It contains AA Cys 24 - Glu 540 (Accession # P42701-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 85.3 kDa. The protein migrates as 100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-12B, His Tag (Cat. No. NK2-H52H7) at 5 μg/mL (100 μL/well) can bind Biotinylated Human IL-12 R beta 1, Fc,Avitag (Cat. No. ILB-H82F7) with a linear range of 10-78 ng/mL (QC tested).;Immobilized Human IL-12B, His Tag (Cat. No. NK2-H52H7) at 2 μg/mL (100 μL/well), can bind pre-mixed increasing concentrations of Anti-IL23A & IL12B MAb (P40 domain) and 0.4 μg/mL (100 μL/well) Biotinylated Human IL-12 R beta 1, Fc,Avitag (Cat. No. ILB-H82F7) with a half maximal inhibitory concentration(IC50) of 0.83 μg/mL (Routinely tested).
Catalog No. 50-195-6875
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.