Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL-13 Monoclonal Antibody (JES10-5A2)

Catalog No. AHC0132
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AHC0132 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AHC0132 Supplier Invitrogen™ Supplier No. AHC0132
Add to Cart
Edge
Add to Cart

Rat Monoclonal Antibody

IL-13 Monoclonal Antibody for Western Blot, IHC, Flow, ELISA, Neu

IL-13 is an immunoregulatory cytokine produced primarily by activated Th2 cells, and is involved in regulating inflammatory and immune responses. IL-13 down-regulates macrophage activity, thereby inhibiting the production of pro-inflammatory cytokines and chemokines, and is involved in allergen-induced asthma through mechanisms independent of IgE and eosinophils. IL-13 also regulates several stages of B cell maturation and differentiation, up-regulates CD23 and MHC class II expression, and promotes IgE isotype switching of B cells. Structurally, IL-13 closely resembles IL-3, IL-4, IL-5 and GM-CSF. The gene encoding IL-13, IL3, IL5, IL4, and CSF2 form a cytokine gene cluster on chromosome 5q, with the IL-13 gene particularly close to IL4.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-13
Applications ELISA, Flow Cytometry, Immunohistochemistry, Neutralization, Western Blot
Classification Monoclonal
Clone JES10-5A2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Il13
Gene Accession No. P35225
Gene Alias ALRH; ALRH antibody; BHR1; il 13; Il13; Il-13; IL-13; cytokine; ILN; Interleukin; interleukin 13; Interleukin13; interleukin-13; interleukin-13 precursor; L13; MGC116786; MGC116788; MGC116789; NC30; NC30 antibody; P600; P600 antibody; T-cell activation protein P600
Gene Symbols Il13
Host Species Rat
Immunogen Purified recombinant human IL-13.
Purification Method Purified
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3596
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Which antibodies directed to cytokines are suitable for use in immunohistochemistry with paraffin sections?

The following antibodies have been used successfully for IHC with paraffin sections:
Mouse (monoclonal) Anti-Human TGF-?1 (Cat. No. AHG0051), Rat (monoclonal) Anti-Human Interleukin-13 (Cat. No. AHC0132), Rabbit (polyclonal) Anti-Human IL-8 (Cat. No. AHC0881), Rabbit (polyclonal) Anti-Mouse Interferon-? (Cat. No. AMC4034), Goat (polyclonal) Anti-Mouse TNF-? (Cat. No. AMC3012), and Rabbit (polyclonal) Anti-Rat TNF-? (Cat. No. ARC3012).

You can also locate these and other antibodies with our helpful Primary Antibody Selection Guide. This can be accessed by searching "Primary Antibody Selector Tool" on our website. Once you've reached the Selection Guide, you can narrow the search by clicking on your desired links at the top of the Guide.

Do you have a neutralizing monoclonal antibody for human IL-13?

Yes, the IL-13 Monoclonal Antibody (JES10-5A2) (Cat. No. AHC0132), clone ID JES10-5A2, has a neutralizing activity against human IL-13.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.