Learn More
BioVendor, LLC Interleukin-13 Receptor Subunit Alpha-2 (IL-13RA2) Human ELISA, 96 wells
SDP
Supplier:  BioVendor, LLC RGP027R
Encompass
Interleukin-13 Receptor Subunit Alpha-2 (IL-13RA2) Human ELISA, 96 wells. Research use only. Immunoassay kit for in vitro quantification of natural or recombinant human IL-13RA2 in serum, plasma, buffered solutions or cell culture medium. Calibration range: 1565000 pg/ml. Detection limit: < 150 pg/ml. Synonyms: IL-13 receptor subunit alpha-2, IL-13R subunit alpha-2, IL-13R-alpha-2, IL13RA2, IL-13R2, Interleukin-13-binding protein, IL-13-binding protein, CD213a2. Summary: IL-13RA2 binds as a monomer with high affinity to IL-13, but not to IL-4. A correlation between the expression level of IL-13RA2 and increased invasiveness as well as metastatic potential of human glioma, breast cancer and pancreatic cancer have been reported. Among normal tissues, IL-13RA2 can be found only in spermatocytes, suggesting that IL-13RA2 can be considered as a good candidate for developing new targeted therapeutics that potentially would exhibit fewer side effects.
Catalog No. 50-206-9238
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.