Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Specifications
Specifications
| Assay Range | 5 - 500 pg/mL |
| Product Type | ELISA Development Kit |
| Sample Type | Serum, Plasma, Cell Culture Supernatant |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 3605 |
| Gene Symbol | IL17A |
| Quantity | 1 Kit (for 6 Plates) |
| Research Discipline | Apoptosis, Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity |
| Storage Requirements | Storage of components varies. See protocol for specific instructions. |
| Synonym | CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, IL-17Acytotoxic T-lymphocyte-associated protein 8, IL-17CTLA-8, IL17interleukin-17A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8), interleukin 17A |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
