Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ IL-17/IL-17A ELISA Development Kit
SDP

Catalog No. DDXKEIL17A Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DDXKEIL17A 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DDXKEIL17A Supplier Novus Biologicals™ Supplier No. DDXKEIL17A
Add to Cart
Add to Cart

Applications: ELISA

The human IL17 ELISA kit is for quantitative determination of IL17 concentrations in human cell culture supernatants and biological fluids (plasma, serum). This kit is for research purpose only. Kit contents: Capture Antibody: 0.5mg/mL of mouse anti-IL17A monoclonal antibody (clone 408H6.01). Dilute in 50mM carbonate buffer pH9.6 to a working concentration of 2.5ug/mL Detection Antibody: 0.5mg/mL of HRPO-conjugated anti-IL17A mouse monoclonal antibody (clone 405B4.01). Dilute to a working concentration of 3ug/mL in PBS-1% BSA-0.05% Tween20 Standard: Each vial contains 1ug/mL of recombinant IL17A produced, purified and concentrated from eukaryotic cells. A 7-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 20ng/mL is recommended.

Specifications

Assay Sensitivity The minimum detectable dose of human Interleukin-17 was determined to be approximately 1ng/mL
Product Type ELISA Kit
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 3605
Gene Symbol IL17A
Kit Contents Capture Antibody: 0.5mg/mL of mouse anti-IL17 monoclonal antibody (clone 408H6.01). Dilute in 50mM carbonate buffer pH 9.6 to a working concentration of 2.5 ug /mL, Detection Antibody: 0.5mg/mL of HRPO-conjugated anti-IL17 mouse monoclonal antibody (clone 405B4.01). Dilute to a working concentration of 3ug/mL in PBS-BSA 1%- Tween 0.05%, Standard: Each vial contains 1ug/mL of recombinant IL17 produced, purified and concentrated from eukaryotic cells. A 7-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 20ng/mL is recommended
Quantity 1 Kit
Research Discipline Apoptosis, Cytokine Research, Immunology, Innate Immunity
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, IL-17Acytotoxic T-lymphocyte-associated protein 8, IL-17CTLA-8, IL17interleukin-17A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8
Target IL-17/IL-17A
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.