Learn More
Medchemexpress LLC IL-17 MODULATOR 3 5MG
SDP
Supplier:  Medchemexpress LLC HY1392035MG
Encompass_Preferred
HY-139203 IL-17 modulator 3 (5 mg). CAS#2467731-88-8. Purity: 99.53%. IL-17 modulator 3 can be used for the research of inflammation, cancer and autoimmune diseases.
Catalog No. NC2596627
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.