Learn More
Medchemexpress LLC Il-17a modulator-1 | 2748749-29-1 | 98.8% | 561.63 g·mol⁻¹ | C33H31N5O4 | 5 MG
Supplier: Medchemexpress LLC HY1454305MG
IL-17A modulator-1 is a small-molecule research compound that inhibits interleukin-17A (IL-17A) signaling. Reported in patent WO2021239743 (example 9), the compound exhibits potent IL-17A inhibitory activity (pIC50 ≈ 8.2) and is intended for in vitro and preclinical studies of immune-mediated and autoimmune diseases, oncology, and neurodegenerative disorder models.
- Potent IL-17A inhibitory activity (pIC50 ≈ 8.2).
- Suitable for in vitro and preclinical research applications.
- High reported purity for consistent experimental results.
- Available in multiple small-molecule pack sizes for dose-ranging studies.
- Provided with analytical data (COA, LCMS, 1H NMR) for quality control.