Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL-17F Monoclonal Antibody (SHLR17), Brilliant Violet™ 786, eBioscience™
GREENER_CHOICE

Catalog No. 417716942
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-771-6942 100 Tests
41-771-6941 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 41-771-6942 Supplier Invitrogen™ Supplier No. 417716942
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rat Monoclonal Antibody

The SHLR17 monoclonal antibody reacts with human interleukin (IL)-17F. IL-17F is a homodimeric, 34 kDa member of the IL-17 family, which includes IL-17(A), B, C, D, E (IL-25). Within this family, IL-17A and IL-17F are produced preferentially by T helper 17 (Th17) cells, with other members produced more widely. IL-17F and IL-17A have many overlapping functions, but knockout studies have shown each to have independent functions as well. In vitro cultured Th17 cells have been shown to express IL-17F and IL-17A homodimers and IL-17A/F heterodimers depending on the culture conditions and their differentiation state. A heterodimer of IL-17RA and IL-17RC has been shown to bind IL-17F, resulting in target cell-secretion of pro-inflammatory cytokines and chemokines, as well as neutrophil recruitment. Applications Tested This SHLR17 antibody has been pre-diluted and tested by intracellular staining followed by flow cytometric analysis of normal human peripheral blood cells using the Intracellular Fixation & Permeabilization Buffer Set (Product # 88-8824-00) and protocol. Please refer to 'Staining Intracellular Antigens for Flow Cytometry, Protocol A: Two step protocol for intracellular (cytoplasmic) proteins' located at Flow Protocols. This may be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can...

IL-17F (Interleukin 17F, CTLA-8)) is a cytokine belonging to the IL-17 family that is produced by inflammatory cells such as activated T cells, mast cells, and basophils. IL-17F is involved in allergic airway inflammation, and can induce several cytokines, chemokines, and adhesion molecules in bronchial epithelial cells, vein endothelial cells, fibroblasts, and eosinophils. IL-17F may be secreted as a homodimer, or a heterodimer with IL17A. It acts by binding to the type I receptor, IL-17R, aiding recruitment of monocytes and neutrophils at the site of inflammation by increasing chemokine production. IL-17F also stimulates induction of other pro-inflammatory cytokines TNF alpha, IL-1 beta, IL-6, and IL-8, and reports strongly suggest the involvement of IL-17 in several chronic inflammatory diseases such as rheumatoid arthritis, psoriasis and multiple sclerosis. TGF-beta (differentiation) and IL-23 (expansion) are required for induction and maintenance of Th17 (IL-17 producing) cells, which in turn induce the other pro-inflammatory cytokines. IL-17F is produced, and exists, as a homo-dimer, with homology to a herpes virus early protein, is one of the six members (IL-17A-F) of this cytokine family, and is well characterized and highly expressed by activated effector memory T cells. IL-17F has been found to inhibit the angiogenesis of endothelial cells and induce endothelial cells to produce IL2, TGFB1/TGFB, and monocyte chemoattractant protein-1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-17F
Applications Flow Cytometry
Classification Monoclonal
Clone SHLR17
Concentration 5 μL/Test
Conjugate Brilliant Violet 786
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene IL17F
Gene Accession No. Q96PD4
Gene Alias C87042; CANDF6; cytokine ML-1; IL17F; IL-17F; IL-24; ILN; Interleukin; interleukin 17F; Interleukin17F; interleukin-17F; interleukin-24; ML1; ML-1; mutant IL-17F
Gene Symbols IL17F
Host Species Rat
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 112744
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.