Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ IL-17RD/SEF Antibody Pair [HRP]

Polyclonal/Monoclonal Antibody Pair
$1195.00
Specifications
Assay Range | 78.125 pg/mL |
---|---|
Assay Sensitivity | 78.13-5000 pg/mL |
Assay | Sandwich ELISA |
Conjugate | HRP |
Product Type | Antibody Pairs |
Description
This solid phase sandwich ELISA utilizes an antibody specific for Human IL-17RD/SEF.Specifications
78.125 pg/mL | |
Sandwich ELISA | |
Antibody Pairs | |
Human | |
FLJ35755, IL-17RDIL-17 receptor D, IL17Rhom, IL17RLMDKFZp434N1928, interleukin 17 receptor D, interleukin-17 receptor D, Interleukin-17 receptor-like protein, MGC133309, Sef homolog, SEFhSef | |
IL17RD | |
Storage is content dependent. |
78.13-5000 pg/mL | |
HRP | |
This solid phase sandwich ELISA utilizes an antibody specific for Human IL-17RD/SEF. | |
Sandwich ELISA | |
54756 | |
Rabbit | |
RUO | |
IL-17RD/SEF |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title