Learn More
MBL International Corporation Mouse IL-18 ELISA Kit
SDP
Supplier:  MBL International Corporation 7625
Encompass_Preferred
Size:96 Wells; The Mouse Il-18 ELISA Kit is based on sandwich ELISA and is capable of measuring mouse Il-18.This kit has a high sensitivity (25.0 pg/ml) and numerous citations.The Mouse IL-18 ELISA Kit is based on sandwich ELISA and capable of measuring mouse IL-18.
Catalog No. 50-446-37
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.