Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL-2 Monoclonal Antibody (419A7A3)
GREENER_CHOICE

Catalog No. AHC0422
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AHC0422 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AHC0422 Supplier Invitrogen™ Supplier No. AHC0422
Season of Savings
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

IL-2 Monoclonal Antibody (419A 7A3)

IL-2 Monoclonal Antibody for ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-2
Applications ELISA
Classification Monoclonal
Clone 419A7A3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.5mg/mL sodium azide; pH 7.4
Gene IL2
Gene Accession No. P60568
Gene Alias aldesleukin; cytokine; H-IL-2; il 2; Il2; Il-2; ILN; Interleukin; interleukin 2; interleukin 2 precursor; interleukin 2 variant IL2delta2; Interleukin2; Interleukin-2; interleukin-2 precursor; interleukin-2 precursor protein; interleukin-2; interleukin 2; Involved in regulation of T cell clonal expansion; involved in regulation of T-cell clonal expansion; lymphokine; M-IL-2; patial sequence of Interleukin-2; POIL2; porcine Interleukin 2; T cell growth factor; T-cell growth factor; T-cell growth factor (TCGF); TCGF
Gene Symbols IL2
Host Species Mouse
Immunogen Purified recombinant human IL-2.
Purification Method Protein A
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3558
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.