Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-2 Rat anti-Mouse, APC, Clone: JES6-5H4, BD

Catalog No. p-7092756 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562041 25 μg
BDB554429 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB562041 Supplier BD Biosciences Supplier No. 562041
Only null left
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The JES6-5H4 monoclonal antibody specifically binds to mouse interleukin-2 (IL-2). IL-2 is a multifunctional cytokine that plays pivotal roles in immunity and tolerance. It is produced by activated T cells. IL-2 effects the activation, growth, proliferation and/or differentiation of various cell types including T and B lymphocytes and their precursors, LAK cells, NK cells, and monocytes/macrophages. IL-2 mediates its biological activities by binding to IL-2 receptor compleses. The intermediate affinity IL-2R is comprised of IL-2R β (CD122) and common gamma chain ( γc; CD132) subunits whereas the high-affinity IL-2R is comprised of IL-2R α (CD25), IL-2R β and γc subunits. The JES6-5H4 monoclonal antibody binds to IL-2 and neutralizes its biological activity.

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-2
Applications Flow Cytometry
Classification Monoclonal
Clone JES6-5H4
Concentration 0.2mg/mL
Conjugate APC
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse IL-2 Recombinant Protein
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.