Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL-2 Monoclonal Antibody (JES6-1A12), Functional Grade, eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. p-7069909
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-146-94 50 μg
50-112-8732 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-146-94 Supplier Invitrogen™ Supplier No. 16702281
Add to Cart
Edge
Add to Cart

Rat Monoclonal Antibody

Description: The JES6-1A12 antibody reacts with mouse interleukin-2 (IL-2). The JES6-1A12 antibody is a neutralizing antibody. Mouse IL-2 (also known as TCGF) is a 17 kDa factor produced mainly by activated CD4+ T cells. IL-2 induces cell cycle progression of resting cells in an antigen non-specific manner and allows clonal expansion of activated T cells. IL-2 also acts on activated B cells, monocytes, NK cells, LAK cells, and on oligodendroglial cells in vitro. In addition, IL-2 plays a role in hematopoiesis, tumor surveillance and anti-inflammatory reactions, and hence is a central regulator of the immune response. Non-glycosylated IL-2 is biologically active. Applications Reported: The JES6-1A12 antibody has been reported for use in ELISA capture, ELISPOT capture, neutralization of IL-2 bioactivity, and IP. Applications Tested: The Functional Grade Purified JES6-1A12 antibody has been tested by LAL assay to verify low endotoxin levels, by ELISA, and in bioassay for neutralization of mouse IL-2 bioactivity. The JES6-1A12 antibody at 0.250 μg/mL has been found to inhibit by 50% the biological effects of 1.0 ng/mL mouse IL-2 in a CTLL-2 cell proliferation assay. Detailed information and protocols about cytokine bioassays and in vitro cytokine neutralization using antibodies can be found in the BestProtocols™ section.

Interleukin 2 (IL-2) is an immuno-modulatory cytokine that is important for the proliferation of activated T cells, differentiation of B cells, natural killer cells, monocytes and macrophages. IL-2 signals through the IL-2 receptor (IL-2R), a heterotrimeric protein complex whose gamma chain is also shared by interleukin 4 (IL-4) and interleukin 7 (IL-7). The expression of the IL-2 gene in mature thymocytes is monoallelic, which represents an unusual regulatory mode for controlling the precise expression of a single gene. The targeted disruption of a gene similar to IL-2 in mice leads to an ulcerative colitis-like disease that suggests an essential role of this gene in the immune response to antigenic stimuli.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-2
Applications Functional Assay, Immunoprecipitation, Immunostaining, Neutralization
Classification Monoclonal
Clone JES6-1A12
Concentration 1 mg/mL
Conjugate Functional Grade
Formulation PBS with no preservative; pH 7.2
Gene IL2
Gene Accession No. P04351
Gene Alias aldesleukin; cytokine; H-IL-2; il 2; Il2; Il-2; ILN; Interleukin; interleukin 2; interleukin 2 precursor; interleukin 2 variant IL2delta2; Interleukin2; Interleukin-2; interleukin-2 precursor; interleukin-2 precursor protein; interleukin-2; interleukin 2; Involved in regulation of T cell clonal expansion; involved in regulation of T-cell clonal expansion; lymphokine; M-IL-2; patial sequence of Interleukin-2; POIL2; porcine Interleukin 2; T cell growth factor; T-cell growth factor; T-cell growth factor (TCGF); TCGF
Gene Symbols IL2
Host Species Rat
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16183
Target Species Mouse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.