Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-20 R beta/FNDC6 Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB320734
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB320734 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB320734 Supplier R&D Systems Supplier No. NBP298428MFV610

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

IL-20 R beta/FNDC6 Polyclonal specifically detects IL-20 R beta/FNDC6 in Rat samples. It is validated for ELISA/Western Blot.

Specifications

Antigen IL-20 R beta/FNDC6
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias FNDC6, DIRS1, IL-20R2, IL20RB interleukin 20 receptor beta, MGC34923
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Rat IL-20 R beta/FNDC6 (Uniprot#: B8K1X9; Met1-Pro227).
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53833
Target Species Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.