Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL-20RB Monoclonal Antibody (20RNTC), eBioscience™
GREENER_CHOICE

Catalog No. 5012839
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
2 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-128-39 100 μg
14-120-637 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-128-39 Supplier Invitrogen™ Supplier No. 14120682
  • $321.00
    $240.65 / Each of 1

    Save $80.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rat Monoclonal Antibody

Description: This 20RNTC monoclonal antibody recognizes mouse and human IL-20R2 (interleukin-20 receptor 2, IL-20Rb) in either of its two heterodimeric complexes formed by pairing with IL-20R1 or IL-22R1. IL-20R2 belongs to the class II cytokine receptor family that shares a characteristic pattern of cysteine, proline and tryptophan residues in the extracellular domain. IL-20R2 paired with IL-20R1 forms a receptor that binds IL-20, IL-19, or IL-24. IL-20R2 paired with IL22R1 forms a receptor that binds IL-20 or IL-24. The IL-20R complex signals primarily via the JAK-STAT pathway. IL-20R2 is found predominantly on cells of epithelial origin such as gut and skin and can be upregulated during inflammation. Immunofluorescent staining of A375 cells reveals a punctate staining pattern at the cell membrane, as well as diffuse, cytoplasmic staining that is likely associated with protein synthesis and transport in the endoplasmic reticulum and golgi. Applications Reported: This 20RNTC antibody has been reported for use in immunocytochemistry, immunoblotting (WB) and flow cytometric analysis. Applications Tested: This 20RNTC antibody has been tested by immunofluorescent staining and flow cytometric analysis of fixed and permeabilized A375 cells. This can be used between 1-10 μg/mL. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.

IL20RB and IL20RA (MIM 605620) form a heterodimeric receptor for interleukin-20 (IL20; MIM 605619) (Blumberg et al., 2001 [PubMed 11163236]).[supplied by OMIM, Feb 2009].
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-20RB
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 20RNTC
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Il20rb
Gene Accession No. Q6UXL0
Gene Alias AV228068; DIRS1; Fibronectin type III domain containing 6; FNDC6; Gm186; IL-20 receptor subunit beta; IL20R2; IL-20R2; IL20RB; IL-20RB; IL-20R-beta; interleukin 20 receptor beta; interleukin 20 receptor beta subunit; interleukin 20 receptor subunit beta; interleukin-20 receptor II; interleukin-20 receptor subunit beta; RGD1566151; UNQ557/PRO1114
Gene Symbols Il20rb
Host Species Rat
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 213208, 53833
Target Species Human, Mouse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.