Learn More
Enzo Life Sciences IL-21 (human), (recombinant) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZPRT2480100
Encompass_Preferred
Alternative name: Interleukin-21. Purity: ≥95% (SDS-PAGE). Formulation: Lyophilized from a sterile (0.2 micron) filtered solution containing 10 mM sodium phosphate, pH 7.5. Source: Produced in E. coli. Contains 132 amino acids. Long Term Storage: -80°C.
Catalog No. 50-201-4061
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.