Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ IL-22 Antibody Pair [HRP]
SDP

Catalog No. NB009851 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
15 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB009851 15 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB009851 Supplier Novus Biologicals™ Supplier No. NBP27940615PLATES
Add to Cart
Add to Cart

Monoclonal Antibody Pair

specifically detects in Human samples. It is validated for ELISA, Sandwich ELISA.

Specifications

Assay Range 2.03 pg/mL
Assay Sensitivity 2.03-130 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human IL-22.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias Cytokine Zcyto18, IL-10-related T-cell-derived inducible factor, IL-21, IL-22IL21, IL-D110, ILTIFIL-10-related T-cell-derived-inducible factor, IL-TIFMGC79382, interleukin 21, interleukin 22, interleukin-22, MGC79384, TIFa, TIFIL-23, zcyto18
Gene ID (Entrez) 50616
Gene Symbol IL22
Host Species Rabbit/Mouse
Kit Contents
  • Standard: Each vial contains 8.8 ng of recombinant Human IL-22.
  • Capture Antibody: 1 mg/mL of mouse anti-Human IL-22 monoclonal antibody (in PBS, pH 7.4).
  • Detection Antibody: 0.2 mg/mL of rabbit anti-Human IL-22 monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4C).
  • Quantity 15 Plates
    Regulatory Status RUO
    Research Discipline Biologically Active Proteins, Cytokine Research, Innate Immunity
    Storage Requirements Storage is content dependent.
    Target IL-22
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 130 pg/mL is recommended.
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.