Learn More
Invitrogen™ IL-23 p19 Chimeric Recombinant Human Monoclonal Antibody (mh066)

Description
Specificity: Human IL23A and Human IL12B Heterodimer. This product is a chimeric monoclonal antibody combining the constant domains of the human IgG1 molecule with mouse variable regions. The antibody was produced using recombinant antibody technology.
Specifications
Specifications
| Antigen | IL-23 p19 Chimeric |
| Applications | Neutralization |
| Classification | Recombinant Monoclonal |
| Clone | mh066 |
| Concentration | 2.25 mg/mL |
| Conjugate | Unconjugated |
| Formulation | arginine, histidine with 120mM NaCl, 0.02% Tween 80 and no preservative |
| Gene | IL23A |
| Gene Accession No. | Q9EQ14, Q9NPF7 |
| Gene Alias | il 23; IL 23 A; IL12B; Il-12b; Il12p40; Il-12p40; il23; IL-23; IL-23 subunit alpha; IL23A; IL-23A; IL-23-A; IL23P19; IL-23p19; ILN; Interleukin; interleukin 12B; interleukin 23 p19 subunit; interleukin 23 subunit alpha; interleukin 23, alpha subunit p19; interleukin-23 subunit alpha; Interleukin-23 subunit p19; interleukin-six, G-CSF related factor; JKA3 induced upon T-cell activation; MGC79388; P19; p40; RP23-388G23.1; SGRF; UNQ2498/PRO5798 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.