Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-27 Mouse anti-Human, Clone: 1E1H5, Proteintech
SDP

Catalog No. 501736598 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6598 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6598 Supplier Proteintech Group Inc Supplier No. 661641IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The protein encoded by IL-27 is one of the subunits of a heterodimeric cytokine complex. This protein is related to interleukin 12A (IL12A). It interacts with Epstein-Barr virus induced gene 3 (EBI3), a protein similar to interleukin 12B (IL12B), and forms a complex that has been shown to drive rapid expansion of naive but not memory CD4(+) T cells. The complex is also found to synergize strongly with interleukin 12 to trigger interferon gamma (IFNG) production of naive CD4(+) T cells. The biological effects of this cytokine are mediated by the class I cytokine receptor (WSX1/TCRR).

Specifications

Antigen IL-27
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1E1H5
Concentration 1.31 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene IL27
Gene Accession No. Q8NEV9
Gene Alias IL 27, IL 27 A, IL 27 subunit alpha, IL 27A, IL27, IL27 A, IL27A, IL27p28, IL30, interleukin 27, Interleukin 27 subunit alpha
Gene Symbols IL27
Host Species Mouse
Immunogen IL-27 Fusion Protein Ag21436
Purification Method Protein A
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 246778
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.