Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL-28 (IFN lambda) Monoclonal Antibody (RN28), eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 5013673
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-136-73 50 μg
50-136-74 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-136-73 Supplier Invitrogen™ Supplier No. 14728181
Add to Cart
Edge
Add to Cart

Rat Monoclonal Antibody

Description: This monoclonal RN28 antibody reacts with IL-28 (IFN lambda), a member of the IFN-lambda family which mediates the induction of anti-viral protection in a wide variety of cells. Applications Reported: The RN28 antibody has been reported for use in ELISA capture of mouse IL-28. Applications Tested: This RN28 antibody has been tested as the capture antibody in a sandwich ELISA for analysis of mouse IL-28 in combination with the biotin polyclonal (cat. 13-7282) antibody for detection. A suitable range of concentrations of this antibody for ELISA capture is 1.0-4 μg/mL. A standard curve consisting of doubling dilutions of the recombinant standard over the range of 4000 pg/mL - 30 pg/mL should be included in each ELISA plate. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.

IL28 is a cytokine distantly related to type I interferons and the IL-10 family. This gene, interleukin 28B (IL28B), and interleukin 29 (IL29) are three closely related cytokine genes that form a cytokine gene cluster on a chromosomal region mapped to 19q13. Expression of the cytokines encoded by the three genes can be induced by viral infection. All three cytokines have been shown to interact with a heterodimeric class II cytokine receptor that consists of interleukin 10 receptor, beta (IL10RB) and interleukin 28 receptor, alpha (IL28RA).
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-28 (IFN lambda)
Applications ELISA
Classification Monoclonal
Clone RN28
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Ifnl3
Gene Accession No. Q4VK74, Q8CGK6
Gene Alias Cytokine Zcyto20; cytokine Zcyto22; EG330496; IFL-1; IFN lambda2; Ifnl2; Ifnl3; IFN-lambda2; IFN-lambda-2; IFN-lambda-3; IFN-lambda-4; IL 28; IL 28 A; Il28; Il28a; IL-28A; Il28b; IL-28B; IL28C; IL-28C; ILN; INF-alpha; INF-lambda; interferon alpha; interferon lambda 2; interferon lambda 3; Interferon lambda2; interferon lambda-2; Interferon lambda-3; interferon, lambda 2; interferon, lambda 3; interferon, lambda 4; interferon-lambda 2; interferon-lambda2; Interleukin; interleukin 28; interleukin 28A; interleukin 28A (interferon, lambda 2); interleukin 28B; interleukin-28A; Interleukin-28B; interleukin-28C; ZCYTO20; ZCYTO22
Gene Symbols Ifnl2, Ifnl3
Host Species Rat
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 330496, 338374
Target Species Mouse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.