Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL-32 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA520222
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA520222 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA520222 Supplier Invitrogen™ Supplier No. PA520222
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is Jurkat cell lysate. PA5-20222 can be used with blocking peptide PEP-0335.

Interleukin-32 (IL-32) was initially identified as a transcript (NK4) that is selectively expressed in lymphocytes and NK cells and whose expression is increased following activation by IL-2. It was later re-isolated from an IL-18-treated lung carcinoma cell line and re-named IL-32. IL-32 is unusual in that it does not share sequence homology with known cytokine families and is highly expressed in immune tissues, existing in at least four differentially spliced isoforms. Because treatment of human monocytic and mouse macrophage cells with IL-32 induces several proinflammatory cytokines such as TNF-a, IL-8 and MIP-2, and because it is also induced in human peripheral lymphocyte cells after mitogen stimulation and in epithelial cells by IFN-gamma, it has been suggested that IL-32 may play a role in autoimmune and inflammatory diseases such as rheumatoid arthritis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-32
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene IL32
Gene Accession No. P24001
Gene Alias IL32; IL-32; IL-32alpha; IL-32beta; IL-32delta; IL-32gamma; ILN; Interleukin; interleukin 32; interleukin-32; interleukin-32 eta; interleukin-32 small; interleukin-32 theta; natural killer cell transcript 4; natural killer cells protein 4; NK4; TAIF; TAIFa; TAIFb; TAIFc; TAIFd; Tumor necrosis factor alpha-inducing factor
Gene Symbols IL32
Host Species Rabbit
Immunogen A 16 amino acid peptide from near the center of human IL-32.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9235
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.