Learn More
AdipoGen anti-IL-33 (human), mAb (IL33305B) (100 µg)
SDP
Supplier:  AdipoGen AG20A0041C100
Encompass_Preferred
Monoclonal Antibody. Recognizes human IL-33. Does not cross-react with mouse IL-33. Isotype: Mouse IgG2akappa. Clone: IL33305B. Applications: ELISA, FUNC (Blocking), IHC, IP, WB. Liquid. 0.2µm-filtered solution in PBS, pH 7.4. Interleukin-33 (IL-33; HF-NEV; IL-1F11), a member of the IL-1 family of cytokines, is expressed by many cell types following pro-inflammatory stimulation and is thought to be released on cell lysis. IL33 binds to and signals through ST2 (IL1R1) and its stimulation recruits MYD88, IRAK, IRAK4, and TRAF6, followed by phosphorylation of ERK1 (MAPK3)/ERK2 (MAPK1), p38(MAPK14), and JNK. The ability of IL-33 to target numerous immune cell types, like Th2-like cells, mast cells, and B1 cells, and to induce cytokine and chemokine production underlines its potential in influencing the outcome of a wide range of diseases, such as arthritis, asthma, atopic allergy & anaphylaxis, cardiovascular disease/atherosclerosis, nervous system diseases, and sepsis.
Catalog No. 50-205-2835
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.