Learn More
Enzo Life Sciences IL-33 (mouse), (recombinant) (10µg)
SDP
Supplier:  Enzo Life Sciences ALX5221013010
Encompass_Preferred
Alternative name: Interleukin-33, NF-HEV, IL-1F11. Concentration: 0.1mg/ml after reconstitution. Purity: ≥95% (SDS-PAGE). Formulation: Lyophilized. Contains PBS. Source: Produced in E. coli. Untagged mouse IL-33 (aa 109-266). Specificity: Binds to mouse and human ST2. Long Term Storage: -20°C.
Catalog No. 50-200-9420
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.