Learn More
AdipoGen IL-38 (aa 3-152) (mouse) (rec.) (FLAG) (10 µg)
SDP
Supplier:  AdipoGen AG40B0101C010
Encompass_Preferred
Protein. Mouse IL-38 [IL-1F10] (aa 3-152) is fused at the C-terminus to a FLAG-tag. Source: E. coli. Endotoxin content: <0.01EU/µg purified protein (LAL test; Lonza). Lyophilized. Contains PBS + 0.5% Trehalose. Purity: >95% (SDS-PAGE). IL-38 (IL-1F10) mRNA is expressed in heart, placenta, fetal liver, spleen, thymus and tonsil. It has been reported that removal of the N-terminus domain of the interleukins of the IL-1F family such as IL-1F5 / 6 / 8 or 9 (also called IL-36Ra, IL-36alpha, IL-36beta or IL-36gamma) is important to increase their biological activity. Recently it has been shown that IL-38 is N-terminally processed and secreted by apoptotic cells. Released processed IL-38 (20-152) binds to the receptor Interleukin-1 receptor accessory protein-like 1 (IL1RAPL1; TIGIRR-2) at the surface of macrophages. Processed IL-38-activated IL1RAPL1 reduces the production of IL-6 leading to inflammation attenuation.
Catalog No. 50-205-3166
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.