Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL-4 Monoclonal Antibody (11B11), APC, eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 5015301
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-153-01 100 μg
50-112-3184 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-153-01 Supplier Invitrogen™ Supplier No. 17704182
Only null left
Add to Cart
Edge
Add to Cart

Rat Monoclonal Antibody

Description: The 11B11 antibody reacts with mouse interleukin-4 (IL-4), a 14 kD cytokine. The 11B11 antibody is a neutralizing antibody. Mouse IL-4, also known as BCDF, BCGF-1 and BSF-1, is secreted by activated Th2 and natural killer T cells, and to a lesser extent by Th1 and mast cells. IL-4 is species-specific and promotes proliferation and differentiation of B cells. IL-4, in conjunction with other signals such as CD40, induces Ig isotype switching of B cells and the upregulation of surface MHC class II and CD23. Applications Reported: This 11B11 antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This 11B11 antibody has been tested by intracellular staining and flow cytometric analysis of Th2 polarized and stimulated mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.

Interleukin-4 (IL-4) is a pleiotropic, immune-modulatory cytokine produced by activated T cells, and serves as a ligand for interleukin 4 receptor. IL-4 has been found to have the broadest range of actions. The interleukin 4 receptor also binds to IL13, which may contribute to many overlapping functions of this cytokine and IL13. STAT6, a signal transducer and activator of transcription, has been shown to play an important role in mediating the immune regulatory signal of IL-4. The IL-4 gene, IL-3, IL-5, IL-13, and CSF2 form a cytokine gene cluster on chromosome 5q. In particular, IL-4 is located in close proximity to IL13 on the chromosome. Further, the IL-4 gene, IL13 and IL5 are found to be coordinated by several long-range regulatory elements over a 120 kilobase range on the chromosome. IL-4 delta 2 is a known splice variant of IL-4.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-4
Applications Flow Cytometry
Classification Monoclonal
Clone 11B11
Concentration 0.2 mg/mL
Conjugate APC
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Il4
Gene Accession No. P07750
Gene Alias B cell growth factor 1; B_cell stimulatory factor 1; B-cell growth factor 1; B-cell IgG differentiation factor; B-cell stimulatory factor 1; BCGF1; BCGF-1; binetrakin; BSF1; BSF-1; cytokine; H-IL-4; IGG1 induction factor; IL2; IL4; Il-4; Il4e12; ILN; Interleukin; interleukin 4; interleukin 4 variant 2; interleukin 4 variant IL-4delta2; interleukin 4 variant IL-4delta3; interleukin 4 variant IL-4int2A; interleukin 4 variant IL-4int3A; interleukin 4 variant IL-4int3B; Interleukin4; interleukin-4; interleukin-4 precursor; lymphocyte stimulatory factor 1; MGC79402; M-IL-4; Pitrakinra
Gene Symbols Il4
Host Species Rat
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16189
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.