Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-4 Mouse, anti-Human, R718, Clone: MP4-25D2, BD Horizon™

Catalog No. p-7214622 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB566981 25 Tests
BDB566980 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB566981 Supplier BD Biosciences Supplier No. 566981
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The MP4-25D2 monoclonal antibody specifically binds to Interleukin-4 (IL-4). IL-4 is also known as Lymphocyte stimulatory factor 1, B cell stimulatory factor 1 (BSF-1), or B cell growth factor 1 (BCGF-1). IL-4 is produced by activated T cells, basophils, and mast cells. IL-4 is a multifunctional cytokine and growth factor that affects a variety of different target cell types. IL-4 can costimulate T cell proliferation and survival, as well as help drive Th2-like cell differentiation and effector functions. It costimulates B cell proliferation and survival, and can help B cells differentiate into IgG4- or IgE-producing cells. IL-4 can also diminish the proinflammatory functions of monocytes and macrophages. IL-4 exerts its biological effects by binding with high affinity to cell surface CD124 (IL-4Rα chain). CD124 forms signaling IL-4 receptor complexes with either CD132/common γ chain or CD213a1/IL-13Rα1 to form either Type I or Type II IL-4 Receptor complexes, respectively. The immunogen used to generate the MP4-25D2 hybridoma was purified recombinant human IL-4. This is a neutralizing antibody. The MP4-25D2 antibody has been reported to crossreact with IL-4 from rhesus monkeys. The use of the MP4-25D2 antibody for epitope mapping of human IL-4 has been described.

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-4
Applications Flow Cytometry
Classification Monoclonal
Clone MP4-25D2
Conjugate R718
Host Species Mouse
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.