Learn More
Enzo Life Sciences IL-5 ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ENZKIT1390001
Encompass_Preferred
The IL-5 ELISA kit is a colorimetric sandwich immunoassay kit with results in 3 hours. Alternative name: Interleukin-5, B cell differentiation factor I, T-cell replacing factor, TRF, EDF, Eosinophil differentiation factor. Assay Time: 3 hours. Applications: ELISA, Colorimetric detection . Species reactivity: Human .
Catalog No. 50-201-3834
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.