Learn More
MyBioSource IL-6 elisa kit :: Rat Interleukin 6 ELISA Kit, x96-Strip-Wells-(Competitive)
SDP
Supplier:  MyBioSource MBS7267075X96COMP
Encompass
This assay has high sensitivity and excellent specificity for detection of IL-6. No significant cross-reactivity or interference between IL-6 and analogues was observed. NOTE: Limited by current skills and knowledge, it is impossible for us to complete the cross-reactivity detection between IL-6 and all the analogues, therefore, cross reaction may still exist in some cases.
Catalog No. NC2751191
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.