Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL-6 Receptor Monoclonal Antibody (B-R6)
GREENER_CHOICE

Catalog No. MA180456
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA180456 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA180456 Supplier Invitrogen™ Supplier No. MA180456
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Human CD126 antibody, clone B-R6 recognizes human Interleukin-6 receptor subunit alpha, also known as CD126, IL-6R or Membrane glycoprotein 80.

Interleukin 6 receptor (IL6R), also known as CD126 (Cluster of Differentiation 126), is a type I cytokine receptor. This gene encodes a subunit of the interleukin 6 (IL6) receptor complex. Interleukin 6 is a potent pleiotropic cytokine that regulates cell growth and differentiation and plays an important role in the immune response. The IL6 receptor is a protein complex consisting of this protein and interleukin 6 signal transducer (IL6ST/GP130/IL6-beta), a receptor subunit also shared by many other cytokines. Dysregulated production of IL6 and this receptor are implicated in the pathogenesis of many diseases, such as multiple myeloma, autoimmune diseases and prostate cancer. Alternatively spliced transcript variants encoding distinct isoforms have been reported. A pseudogene of this gene is found on chromosome 9.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-6 Receptor
Applications ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, Immunocytochemistry
Classification Monoclonal
Clone B-R6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with <0.1% sodium azide
Gene IL6R
Gene Accession No. P08887
Gene Alias CD126; CD126 antigen; gp80; IL-6 Receptor; il-6 receptor alpha; IL-6 receptor alpha chain; IL-6 receptor subunit alpha; IL6Q; IL6R; IL-6R; IL-6R 1; IL-6R subunit alpha; IL6R1; IL-6R-1; Il6ra; IL-6RA; IL-6R-alpha; IL6RQ; ILN; Interleukin; interleukin 6 receptor; interleukin 6 receptor, alpha; interleukin-6 receptor subunit alpha; Membrane glycoprotein 80; MGC104991
Gene Symbols IL6R
Host Species Mouse
Immunogen CHO cells transfected with cDNA of the IL-6 receptor.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3570
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.