Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse PGAM1 His Protein
SDP

Catalog No. NBP252614 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP252614 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP252614 Supplier R&D Systems™ Supplier No. NBP252614

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity

A bioactive recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-254 of Mouse PGAM1 The Recombinant Mouse PGAM1 Protein is derived from E. coli. The Recombinant Mouse PGAM1 Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.

Specifications

Concentration 1mg/mL
For Use With (Application) Bioactivity, SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH8.0), 20% glycerol, 0.1 M NaCl, 1mM DTT
Gene ID (Entrez) 5223
Molecular Weight (g/mol) TMW: 31.4kDa
Quantity 100 μg
Source E. coli
Storage Requirements Store at 4°C short term. Aliquot and store at −70°C long term. Avoid freeze-thaw cycles.
Gene Alias BPG-dependent PGAM 1, EC 3.1.3.13, EC 5.4.2.1, EC 5.4.2.4, PGAMA, PGAM-Bphosphoglycerate mutase 1, phosphoglycerate mutase 1 (brain), phosphoglycerate mutase A, nonmuscle form, Phosphoglycerate mutase isozyme B
Common Name PGAM1
Conjugate Unconjugated
Product Line Novus
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.