Learn More
Description
In ELISAs, does not cross-react with bovine FGF acidic, recombinant human (rh) G-CSF, rhGM-CSF, rhIL-1 alpha, rhIL-1 beta, rhIL-2, rhIL-3, rhIL-4, rhIL-6, recombinant mouse IL-7, rhIL-8, porcine PDGF, rhTNF-alpha, rhTNF-beta, human TGF-beta 1, or porcine TGF-beta 1. Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
Specifications
Specifications
| Antigen | IL-7 |
| Applications | ELISA, Neutralization, Western Blot |
| Classification | Monoclonal |
| Clone | 7417 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative; pH 7.4 |
| Gene | Il7 |
| Gene Accession No. | P13232, P16871 |
| Gene Alias | A630026I06Rik; H-IL-7; hlb368; IL7; Il-7; ILN; Interleukin; interleukin 7; Interleukin7; interleukin-7; M-IL-7 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
