Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL-8 Human Matched Antibody Pair

Catalog No. 5017943
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-179-43 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-179-43 Supplier Invitrogen™ Supplier No. BMS2043MST

missing translation for 'validMainframeMsg'

Includes

  • 1 vial (5.5mL) monoclonal Coating Antibody to human IL-8/NAP-1 (100μg/mL)
  • 1 vial (55μL) Biotin-Conjugate anti-human IL-8/NAP-1 polyclonalantibody
  • 1 vial (11μL) Streptavidin-HRP
  • 1 vial (30μL) human IL-8/NAP-1 Standard protein, 200ng/mL
  • 2 vials (50mL) Sample Diluent

Matched Antibody Pair

The Human IL-8/NAP-1 Matched Antibody Pair Kit comes with pre-matched antibody pairs,standards,and streptavidin-HRP to develop your own enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of human IL-8. Buffer reagents and microplates needed to complete the ELISA reaction are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate,which is an overnight process. Samples,standards,or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody,a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.

IL-8 (Interleukin-8) is a 72 amino acid pro-inflammatory CXC chemokine designated CXCL8,secreted by endothelial cells and monocytes,which acts as a major chemoattractant for neutrophils and also T cells and basophils. IL-8,first isolated from osteosarcoma cells,contains the ELR-motif (N-terminal Glu-Leu-Arg amino acid sequence) and signals through the CXCR1 and CXCR2 receptors. Cancer studies have demonstrated a role for IL-8 in the angiogenesis and growth of tumours. Previous nomenclature for IL-8 includes neutrophil activating protein 1 (NAP-1),granulocyte chemotactic protein 1 (GCP-1),monocyte-derived neutrophil-activating peptide (MONAP) and protein 3-10C.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-8
Applications ELISA
Classification Monoclonal/Polyclonal
Conjugate Biotin
Gene Accession No. P10145
Gene Alias Protein 3-10C, CXCL8, NAP-1, GCP-1
Gene Symbols CXCL8
Quantity 10 x 96 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3576
Target Species Human
Content And Storage -20°C
Product Type ELISA Kit
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.