Learn More
Carbosynth LLC. Purified recombinant Human IL1 beta protein
SDP
Supplier:  Carbosynth LLC. 30RAI003
Encompass_Preferred
IL1beta is a proinflammatory cytokine produced in a variety of cells including monocytes, tissue macrophages, keratinocytes and other epithelial cells. Both IL1a and IL1beta binds to the same receptor and has similar if not identical biological properties. These cytokines have a broad range of activities including, stimulation of thymocyte proliferation, by inducing IL2 release, B-cell maturation and proliferation, mitogenic FGFlike activity and the ability to stimulate the release of prostaglandin and collagenase from synovial cells. However, whereas IL1beta is a secreted cytokine, IL1a is predominantly a cell-associated cytokine.
Catalog No. 50-125-3207
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.