Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL12RB2 Recombinant Rabbit Monoclonal Antibody (093)
GREENER_CHOICE

Catalog No. PIMA531162 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531162 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531162 Supplier Invitrogen™ Supplier No. MA531162
Add to Cart
Edge
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Mouse IL12RB2.

The protein encoded by this gene is a type I transmembrane protein identified as a subunit of the interleukin 12 receptor complex. The coexpression of this and IL12RB1 proteins was shown to lead to the formation of high-affinity IL12 binding sites and reconstitution of IL12 dependent signaling. The expression of this gene is up-regulated by interferon gamma in Th1 cells, and plays a role in Th1 cell differentiation. The up-regulation of this gene is found to be associated with a number of infectious diseases, such as Crohn's disease and leprosy, which is thought to contribute to the inflammatory response and host defense.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL12RB2
Applications ELISA
Classification Recombinant Monoclonal
Clone 093
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene IL12RB2
Gene Accession No. P97378
Gene Alias A930027I18Rik; Ifnm; IL-12 receptor subunit beta-2; IL-12R subunit beta-2; Il12rb2; IL-12RB2; IL-12R-beta-2; interferon response to microorganisms; interleukin 12 receptor subunit beta 2; interleukin 12 receptor, beta 2; interleukin-12 receptor beta-2 chain; interleukin-12 receptor subunit beta-2; RP11-102M16.1
Gene Symbols IL12RB2
Host Species Rabbit
Immunogen Recombinant Mouse IL12RB2 protein (Met1-Asn637).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16162
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.