Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL15RA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA579467 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA579467 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA579467 Supplier Invitrogen™ Supplier No. PA579467
Season of Savings
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human placenta tissue. IHC: mouse brain tissue, rat brain tissue. ICC/IF: K562 cell. Flow: K562 cell.

Interleukin 15 (IL-15RA) is a cytokine receptor that specifically binds IL-15 with high affinity. IL-15 regulates T and natural killer cell activation and proliferation. The IL-15 and IL-2 receptors share two subunits, IL-2R beta and IL-2R gamma, and IL-15RA is structurally related to IL-2RA, an additional IL-2-specific alpha subunit necessary for high affinity IL-2 binding. Unlike IL-2RA, IL-15RA is capable of binding IL-15 with high affinity independent of the other subunits. IL-15RA is thought to enhance cell proliferation and expression of apoptosis inhibitor Bcl-xL and Bcl-2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL15RA
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation Antibody with 4mg trehalose, 0.9mg NaCl, 0.2mg Na2PO4 and no preservative
Gene IL15RA
Gene Accession No. Q13261, Q60819
Gene Alias AA690181; CD215; IL 15 receptor; IL 15 receptor subunit α; IL 15R; IL 15R alpha; IL 15R α; IL15 receptor; IL15 receptor subunit alpha; IL-15 receptor subunit alpha; IL15 receptor subunit α; IL15R; IL15R alpha; IL15R α; Il15ra; IL-15RA; IL-15R-alpha; Interleukin 15 receptor; interleukin 15 receptor alpha isoform EM2; interleukin 15 receptor alpha isoform IC2; interleukin 15 receptor alpha isoform IC3; interleukin 15 receptor alpha isoform IC4; interleukin 15 receptor alpha isoform IC5; interleukin 15 receptor alpha isoform IC6; interleukin 15 receptor alpha isoform IC7; interleukin 15 receptor alpha isoform IC8; interleukin 15 receptor subunit alpha; Interleukin 15 receptor subunit α; interleukin 15 receptor, alpha; interleukin 15 receptor, alpha chain; interleukin-15 receptor subunit alpha; sIL 15R; sIL-15 receptor subunit alpha; sIL15R; sIL-15RA; sIL-15R-alpha; soluble IL 15R; soluble IL15R; Soluble interleukin-15 receptor subunit alpha
Gene Symbols IL15RA
Host Species Rabbit
Immunogen E. coli-derived human IL15RA recombinant protein (Position: I31-T174).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16169, 3601, 690369
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.