Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL17B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595491
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595491 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595491 Supplier Invitrogen™ Supplier No. PA595491
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: mouse NIH3T3 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

The protein encoded by this gene is a T cell-derived cytokine that shares sequence similarity with IL17. This cytokine was reported to stimulate the release of TNF alpha (TNF) and IL1 beta (IL1B) from a monocytic cell line. Immunohistochemical analysis of several nerve tissues indicated that this cytokine is primarily localized to neuronal cell bodies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL17B
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and 0.05mg sodium azide
Gene IL17B
Gene Accession No. Q9QXT6, Q9UHF5
Gene Alias 1110006O16Rik; 1700006N07Rik; Cytokine CX1; Cytokine Zcyto7; Cytokine-like protein ZCYTO7; gene for cytokine-like protein; IL17B; IL-17B; IL20; IL-20; Interleukin; interleukin 17B; interleukin 20; interleukin-17 beta; interleukin-17B; Interleukin20; Interleukin-20; neuronal interleukin-17 related factor; neuronal interleukin-17-related factor; NIRF; UNQ516/PRO1031; ZCYTO7
Gene Symbols IL17B
Host Species Rabbit
Immunogen E.coli-derived mouse IL17B recombinant protein (Position: R23-F180).
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116472, 27190, 56069
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.