Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL17RE Monoclonal Antibody (46N7E3)
GREENER_CHOICE

Catalog No. PIMA524806
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524806 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524806 Supplier Invitrogen™ Supplier No. MA524806
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Immunogen sequence: MVHLLVQKSK KSSTFKFYRR HKMPAPAQRK LLPRRHLSEK SHHISIPSPD ISHKGLRSKR TQPSDPETWE SLPRLDSQRH GGPEFSFDLL PEARAIRVTI SSGPE.

The interleukin 17 receptor E (IL-17RE) shares limited similarity to the receptor of IL-17A, a ubiquitous type I membrane glycoprotein that with IL-17A plays a pathogenic role in many inflammatory and autoimmune diseases such as rheumatoid arthritis. IL-17RE is the functional receptor of IL-17C and has been suggested to mediate mucosal immunity to infection with intestinal pathogens. IL-17C is produced by epithelia in response to bacterial challenge and binds to a receptor complex formed by IL-17RE and IL-17RA on the epithelial cell surface, thereby regulating the immune function of epithelial cells in an autocrine manner.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL17RE
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 46N7E3
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA and 0.05% sodium azide
Gene IL17RE
Gene Accession No. Q8NFR9
Gene Alias AA589509; IL-17 receptor E; IL17RE; IL-17RE; Il25r; interleukin 17 receptor E; interleukin-17 receptor E; UNQ3056/PRO9877
Gene Symbols IL17RE
Host Species Mouse
Immunogen Amino acids 97-199.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 132014
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.