Learn More
Description
- Kit is based on sandwich enzyme-linked immune-sorbent assay technology
- A monoclonal antibody from rat-specific for IL-1ß was pre-coated onto 96-well plates
- Store at 4°C for 6 months, at -20°C for 12 months; avoid multiple freeze-thaw cycles
- For research use only
Specifications
Specifications
| Calibration Range | 12.5 to 800pg/mL |
| Description | Il1b (Mouse) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of mouse Il1b. |
| Detection Method | Colorimetric |
| Product Type | Il1b (Mouse) ELISA Kit |
| Sample Type | Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum |
| Target Species | Mouse |
| For Use With (Application) | Quantification |
| Gene ID (Entrez) | 16176 |
| Gene Symbol | Gene Alias: IL-1beta,Il-1b |
| Quantity | 1 Kit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.