Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ IL1R1 (Human) ELISA Kit

Catalog No. 89032270 Shop All Abnova Corporation Products
Change view
Click to view available options
Target Species:
Human
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Target Species
89-032-270 Human
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-032-270 Supplier Abnova™ Supplier No. KA4902
Add to Cart
Add to Cart

Sandwich enzyme immunoassay for the quantitative measurement of human IL1R1

  • For quantitative detection of IL-1R1 in human serum, plasma, cell culture supernatants or tissue homogenate

Specifications

Calibration Range 31.2 to 2000 pg/mL
Description IL1R1 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human IL1R1.
Conjugate Biotin
Detection Method Colorimetric
Product Type ELISA Kit
Sample Type Buffered Solution, Cell Lysates, Cell Culture Supernatant, Plasma, Serum
Target Species Human
For Use With (Application) Quantification
Gene ID (Entrez) 3554
Kit Contents One 96-well plate pre-coated with anti-human IL-1R1 antibody: 96 (8x12) wells, Lyophilized human IL-1R1 standards: 10ng/tube x 2;
Regulatory Status For research use only (RUO)
Sample Volume 100 μL
Storage Requirements Store at 4°C for 4 months, or at -20°C for 8 months.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.