Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL1R2 Mouse anti-Human, Mouse, Clone: 3H4H4, Proteintech
SDP

Catalog No. 501736359 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6359 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6359 Supplier Proteintech Group Inc Supplier No. 602621IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The protein encoded by this gene is a cytokine receptor that belongs to the interleukin 1 receptor family. This protein binds interleukin alpha (IL1A), interleukin beta (IL1B), and interleukin 1 receptor, type I(IL1R1/IL1RA), and acts as a decoy receptor that inhibits the activity of its ligands. Interleukin 4 (IL4) is reported to antagonize the activity of interleukin 1 by inducing the expression and release of this cytokine. This gene and three other genes form a cytokine receptor gene cluster on chromosome 2q12. Two alternatively spliced transcript variants encoding the same protein have been reported.

Specifications

Antigen IL1R2
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 3H4H4
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene IL1R2
Gene Accession No. P27930, P27931
Gene Alias CD121b, CDw121b, IL 1R 2, IL 1R beta, IL 1RT 2, IL 1RT2, IL1 Receptor 2, IL-1 Receptor 2, IL1 Receptor II, IL-1 Receptor II, IL1R2, IL1RB, IL-1RII, Interleukin 1 receptor beta, Interleukin 1 receptor type 2, Interleukin 1 receptor type II
Gene Symbols Il1r2
Host Species Mouse
Immunogen IL1R2 Fusion Protein Ag4495
Purification Method Protein A
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16178, 7850
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.