Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PeproTech IL1RA Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™

Catalog No. 502718610
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-271-8610 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-271-8610 Supplier PeproTech Supplier No. 500P209BT500UG
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AA Sequence of recombinant protein: MRPSGRKSSK MQAFRIWDVN QKTFYLRNNQ LVAGYLQGPN VNLEEKIDVV PIEPHALFLG IHGGKMCLSC VKSGDETRLQ LEAVNITDLS ENRKQDKRFA FIRSDSGPTT SFESAACPGW FLCTAMEADQ PVSLTNMPDE GVMVTKFYFQ EDE. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human IL-1RA. Anti-Human IL-1RA-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Human IL-1RA by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human IL-1RA (500-P209) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human IL-1RA. Western Blot: To detect Human IL-1RA by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. When used in conjunction with compatible secondary reagents the detection limit for Recombinant Human IL-1RA is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P209BT-1MG will be provided as 2 x 500 μg

The protein encoded by this gene is a cytokine receptor that belongs to the interleukin 1 receptor family. This protein is a receptor for interleukin alpha (IL1A), interleukin beta (IL1B), and interleukin 1 receptor, type I(IL1R1/IL1RA). It is an important mediator involved in many cytokine induced immune and inflammatory responses. This gene along with interleukin 1 receptor, type II (IL1R2), interleukin 1 receptor-like 2 (IL1RL2), and interleukin 1 receptor-like 1 (IL1RL1) form a cytokine receptor gene cluster in a region mapped to chromosome 2q12.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL1RA
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.1-1.0 mg/mL
Conjugate Biotin
Formulation PBS with no preservative
Gene IL1RN
Gene Accession No. P18510
Gene Alias Anakinra; CD121A; cytokine; D2S1473; DIRA; F630041P17Rik; ICIL-1RA; IL 1 receptor antagonist; IL1 inhibitor; il1 receptor; il-1 receptor; IL1 receptor antagonist; IL1F3; IL1R; il-1r; IL1RA; IL-1RA; IL1-RA; IL-1ra3; IL-1R-alpha; IL1RN; IL-1RN; IL1RN (IL1F3); interleukin 1 receptor antagonist; interleukin-1 receptor; interleukin-1 receptor antagonist; interleukin-1 receptor antagonist protein; interleukin-1 receptor antagonist secretory form; intracellular IL-1 receptor antagonist type II; intracellular interleukin-1 receptor antagonist (icIL-1ra); IRAP; IRAP1; MGC10430; MVCD4; P80; type II interleukin-1 receptor antagonist
Gene Symbols IL1RN
Host Species Rabbit
Immunogen E.coli-derived Recombinant Human IL-1RA.
Purification Method Antigen affinity chromatography
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3557
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.