Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL1RAPL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547039
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547039 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547039 Supplier Invitrogen™ Supplier No. PA547039
Only null left
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In direct ELISAs, approximately 35% cross-reactivity with recombinant mouse IL-1 RAPL2/IL-1 R9 is observed. In Western blots, approximately 5% cross-reactivity with recombinant human (rh) IL-1 R7 is observed and less than 1% cross-reactivity with rhIL-1 R2 and rhIL-1 R8 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

The protein encoded by this gene is a member of the interleukin 1 receptor family. This protein is similar to the interleukin 1 accessory proteins, and is most closely related to interleukin 1 receptor accessory protein-like 1 (IL1RAPL1). This gene and IL1RAPL1 are located at a region on chromosome X that is associated with X-linked non-syndromic mental retardation. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL1RAPL2
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene IL1RAPL2
Gene Accession No. Q9NP60
Gene Alias IL-1 receptor accessory protein-like 2; IL1R9; IL-1R9; IL-1R-9; IL1RAPL2; IL-1-RAPL-2; IL-1RAPL-2; IL1RAPL-2; IL1RAPL-2-related protein; interleukin 1 receptor 9; interleukin 1 receptor accessory protein like 2; interleukin 1 receptor accessory protein-like 2; Interleukin-1 receptor 9; RGD1561761; three immunoglobulin domain-containing IL-1 receptor-related 1; TIGIRR-1; X-linked interleukin-1 receptor accessory protein-like 2
Gene Symbols IL1RAPL2
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human IL-1 RAPL2/IL-1 R9 Thr17-Glu356.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26280
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.