Learn More
Invitrogen™ IL20RA Polyclonal Antibody

Description
This antibody is predicted to react with chimpanzee and rat samples based on sequence homology. Suggested positive control: A375. The amino acid sequence used as immunogen is 100% homologous in human and chimp, and 82% homologous in mouse and rat.
Specifications
Specifications
| Antigen | IL20RA |
| Applications | Flow Cytometry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.2% gelatin and 0.05% sodium azide |
| Gene | Il20ra |
| Gene Accession No. | Q6PHB0, Q9UHF4 |
| Gene Alias | class II cytokine receptor ZCYTOR7; CRF2-8; Cytokine receptor class-II member 8; Cytokine receptor family 2 member 8; E230031K19; IL-20 receptor subunit alpha; IL-20R1; IL20RA; IL-20RA; IL-20R-alpha; interleukin 20 receptor alpha subunit; interleukin 20 receptor subunit alpha; interleukin 20 receptor, alpha; interleukin-20 receptor I; interleukin-20 receptor subunit alpha; PRO1315; UNQ681; UNQ681/PRO1315; ZcytoR7 |
| Gene Symbols | Il20ra |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.